KEY465193186 Key Organics / BIONET

ethyl 5-cyano-6-{[(2-methoxyphenyl)methyl]amino}-2-phenylpyridine-3-carboxylate

  • MDL number MFCD00793667
  • Cas Number : 477866-14-1
  • Molecular Weight : 387.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64517

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1L-332S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart