KEY465193193 Key Organics / BIONET

5-{[(4-methoxyphenyl)sulfanyl]methylidene}-2,2-dimethyl-1,3-dioxane-4,6-dione

  • MDL number MFCD00794026
  • Cas Number : 477866-31-2
  • Molecular Weight : 294.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64540

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1L-362S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart