KEY465193199 Key Organics / BIONET

5-{[(4-bromophenyl)sulfanyl]methylidene}-2,2-dimethyl-1,3-dioxane-4,6-dione

  • MDL number MFCD00794032
  • Cas Number : 477866-37-8
  • Molecular Weight : 343.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64549

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1L-373S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart