KEY465193221 Key Organics / BIONET

1-methyl-3-{[(4-methylphenyl)sulfanyl]methyl}-1H-pyrazol-5-ol

  • MDL number MFCD00794829
  • Cas Number : 321526-26-5
  • Molecular Weight : 234.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64662

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1L-588S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart