KEY465193227 Key Organics / BIONET

(2Z,4E)-2,3-dichloro-4-{2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]hydrazin-1-ylidene}but-2-enoic acid

  • MDL number MFCD00793840
  • Cas Number : 477867-48-4
  • Molecular Weight : 362.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64725

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1L-920_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart