KEY465193228 Key Organics / BIONET

3-chloro-N-({3-[5-(methylamino)-1,3,4-thiadiazol-2-yl]-4,5-dihydro-1,2-oxazol-5-yl}methyl)-5-(trifluoromethyl)pyridin-2-amine

  • MDL number MFCD01315923
  • Cas Number : 477867-49-5
  • Molecular Weight : 392.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64727

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1L-922_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart