KEY465193230 Key Organics / BIONET

(3E)-3-[2-(2-methylphenyl)hydrazin-1-ylidene]piperidin-2-one

  • MDL number MFCD00794637
  • Cas Number : 14490-03-0
  • Molecular Weight : 217.272

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64729

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1L-925_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart