KEY465193232 Key Organics / BIONET

1-(pyridin-2-yl)-4-{[5-(1,2,3-thiadiazol-4-yl)thiophen-2-yl]sulfonyl}piperazine

  • MDL number MFCD00793739
  • Cas Number : 477867-54-2
  • Molecular Weight : 393.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64734

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1M-006_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart