KEY465193240 Key Organics / BIONET

ethyl 2-{[3-(2-chloro-6-fluorophenyl)-5-methyl-1,2-oxazol-4-yl]formamido}acetate

  • MDL number MFCD00793762
  • Cas Number : 400823-29-2
  • Molecular Weight : 340.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64762

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1M-035_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart