KEY465193241 Key Organics / BIONET

1-tert-butyl-3-(4-fluorophenyl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine

  • MDL number MFCD01568285
  • Cas Number : 338391-69-8
  • Molecular Weight : 285.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64772

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1M-307S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart