KEY465193243 Key Organics / BIONET

(3Z)-3-[2-(4-methylphenyl)hydrazin-1-ylidene]-1-[(morpholin-4-yl)methyl]-2,3-dihydro-1H-indol-2-one

  • MDL number MFCD01314850
  • Cas Number : 1164499-08-4
  • Molecular Weight : 350.422

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64774

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1M-309S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart