KEY465193246 Key Organics / BIONET

(3Z)-1-[(4-methylpiperazin-1-yl)methyl]-3-(2-phenylhydrazin-1-ylidene)-2,3-dihydro-1H-indol-2-one

  • MDL number MFCD01314854
  • Cas Number : 338391-76-7
  • Molecular Weight : 349.438

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64781

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1M-320S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart