KEY465193248 Key Organics / BIONET

6-methyl-3-(4-nitrophenyl)-2H-1,4-benzoxazine

  • MDL number MFCD02185893
  • Cas Number : 400078-33-3
  • Molecular Weight : 268.272

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64785

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1M-326S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart