KEY465193255 Key Organics / BIONET

3-amino-N-(2-methylphenyl)-5-(methylsulfanyl)-4H-1,2,4-triazole-4-carboxamide

  • MDL number MFCD01314903
  • Cas Number : 338391-95-0
  • Molecular Weight : 263.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64823

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1M-535S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart