KEY465193260 Key Organics / BIONET

ethyl 2-(3-chloro-2,2-dimethylpropanamido)-3-phenylpropanoate

  • MDL number MFCD00794137
  • Cas Number : 1396999-17-9
  • Molecular Weight : 311.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64846

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1M-706_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart