KEY465193269 Key Organics / BIONET

(4Z)-4-[2-(4-chloronaphthalen-1-yl)hydrazin-1-ylidene]-2-methyl-1,2,3,4-tetrahydroisoquinoline-1,3-dione

  • MDL number MFCD08064711
  • Cas Number : 924862-22-6
  • Molecular Weight : 363.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64858

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1M-719_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart