KEY465193272 Key Organics / BIONET

5-[(E)-2-(3-chloro-1-benzothiophen-2-yl)ethenyl]-3-methyl-4-nitro-1,2-oxazole

  • MDL number MFCD00794151
  • Cas Number : 477868-37-4
  • Molecular Weight : 320.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64871

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1M-736_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart