KEY465193283 Key Organics / BIONET

1-[(3-methoxypropyl)amino]-3-[(2-methyl-1H-indol-4-yl)oxy]propan-2-ol

  • MDL number MFCD01315614
  • Cas Number : 338392-19-1
  • Molecular Weight : 292.379

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64897

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1M-918_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart