KEY465193288 Key Organics / BIONET

(4E)-2-methyl-6-oxo-4-({[4-(trifluoromethoxy)phenyl]amino}methylidene)-1,4,5,6-tetrahydropyridine-3-carbonitrile

  • MDL number MFCD00974999
  • Cas Number : 338392-24-8
  • Molecular Weight : 323.275

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64906

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1M-931_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart