KEY465193296 Key Organics / BIONET

N-tert-butylmorpholine-4-carbothioamide

  • MDL number MFCD24391363
  • Cas Number : 14294-00-9
  • Molecular Weight : 202.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83 - 86
Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/402876

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1N-506S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart