KEY465193297 Key Organics / BIONET

1-(4-chlorophenyl)-2-[(4-methyl-4H-1,2,4-triazol-3-yl)sulfanyl]ethan-1-one

  • MDL number MFCD00169749
  • Cas Number : 301193-48-6
  • Molecular Weight : 267.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Physical Form Off-white to Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64972

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/1N-510S_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart