KEY465193298 Key Organics / BIONET

4-[(1E)-(methoxyimino)methyl]-1-methyl-3-phenyl-1H-pyrazol-5-ol

  • MDL number MFCD01315295
  • Cas Number : 321533-56-6
  • Molecular Weight : 231.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64973

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1N-511S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart