KEY465193303 Key Organics / BIONET

methyl 1-(4-fluorophenyl)-5-hydroxy-1H-pyrazole-3-carboxylate

  • MDL number MFCD05668743
  • Cas Number : 1007073-76-8
  • Molecular Weight : 236.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65004

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1N-577S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart