KEY465193307 Key Organics / BIONET

(3E,5E)-6-(dimethylamino)-1,1,1-trifluoro-4-[(4-fluorophenyl)amino]hexa-3,5-dien-2-one

  • MDL number MFCD01568310
  • Cas Number : 338393-25-2
  • Molecular Weight : 302.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65017

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1N-721_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart