KEY465193308 Key Organics / BIONET

methyl 2-{[2-(trifluoromethyl)pyridin-4-yl]amino}benzoate

  • MDL number MFCD01568312
  • Cas Number : 338393-28-5
  • Molecular Weight : 296.249

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65019

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1N-723_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart