KEY465193309 Key Organics / BIONET

(3E)-4-[(4-chlorophenyl)amino]-1,1,1-trifluoropent-3-en-2-one

  • MDL number MFCD01315387
  • Cas Number : 338393-32-1
  • Molecular Weight : 263.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Physical Form Off-white to Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65023

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/1N-729_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart