KEY465193311 Key Organics / BIONET

ethyl 1-{5-[(1E)-{[(pyridin-4-yl)formamido]imino}methyl]-1,3-thiazol-2-yl}piperidine-4-carboxylate

  • MDL number MFCD01814587
  • Cas Number : 477868-54-5
  • Molecular Weight : 387.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65047

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1P-015_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart