KEY465193325 Key Organics / BIONET

4-hydroxy-2-oxo-N'-[(1E)-(1H-pyrrol-2-yl)methylidene]-1-azatricyclo[7.3.1.0^{5,13}]trideca-3,5(13),6,8-tetraene-3-carbohydrazide

  • MDL number MFCD01814596
  • Cas Number : 477868-68-1
  • Molecular Weight : 336.351

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65098

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1P-078_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart