KEY465193326 Key Organics / BIONET

4-hydroxy-N'-[(1E)-(3-hydroxyphenyl)methylidene]-2-oxo-1-azatricyclo[7.3.1.0^{5,13}]trideca-3,5(13),6,8-tetraene-3-carbohydrazide

  • MDL number MFCD01814597
  • Cas Number : 477868-69-2
  • Molecular Weight : 363.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65099

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1P-079_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart