KEY465193327 Key Organics / BIONET

N'-[(1E)-[2-(piperidin-1-yl)-1,3-thiazol-5-yl]methylidene]-4-(trifluoromethyl)benzohydrazide

  • MDL number MFCD01814599
  • Cas Number : 477868-71-6
  • Molecular Weight : 382.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65101

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/1P-081_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart