KEY465193333 Key Organics / BIONET

N'-{8-methyl-8H-thieno[2,3-b]indole-2-carbonyl}-2-(thiophen-2-yl)acetohydrazide

  • MDL number MFCD01814710
  • Cas Number : 477869-01-5
  • Molecular Weight : 369.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65155

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1P-377S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart