KEY465193345 Key Organics / BIONET

methyl 3-{2-[(4-bromophenyl)sulfanyl]acetamido}thiophene-2-carboxylate

  • MDL number MFCD01814719
  • Cas Number : 477869-14-0
  • Molecular Weight : 386.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65181

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1P-539S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart