KEY465193347 Key Organics / BIONET

1-({4-[(4-chlorophenyl)sulfanyl]-3-nitrophenyl}methyl)piperidine

  • MDL number MFCD01814722
  • Cas Number : 477869-36-6
  • Molecular Weight : 362.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65209

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1P-647S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart