KEY465193352 Key Organics / BIONET

4-[(3-chlorophenoxy)methyl]-1-[(4-methylphenyl)sulfanyl]-2-nitrobenzene

  • MDL number MFCD01814998
  • Cas Number : 477869-60-6
  • Molecular Weight : 385.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65235

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1P-679S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart