KEY465193353 Key Organics / BIONET

4-[(4-methylphenoxy)methyl]-1-[(4-methylphenyl)sulfanyl]-2-nitrobenzene

  • MDL number MFCD01815000
  • Cas Number : 477869-62-8
  • Molecular Weight : 365.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65237

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1P-681S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart