KEY465193355 Key Organics / BIONET

4-ethyl-5-{4H,5H-naphtho[1,2-b]thiophen-2-yl}-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD01815411
  • Cas Number : 477869-75-3
  • Molecular Weight : 313.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65251

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1P-695S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart