KEY465193360 Key Organics / BIONET

3-[(2-chloro-6-fluorophenyl)methyl]-4-hydroxy-1-[3-(1H-imidazol-1-yl)propyl]-1,2-dihydropyridin-2-one

  • MDL number MFCD02082375
  • Cas Number : 477869-81-1
  • Molecular Weight : 361.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65258

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1P-709_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart