KEY465193361 Key Organics / BIONET

3-[(2-chloro-6-fluorophenyl)methyl]-4-hydroxy-1-[(3-nitropyridin-2-yl)amino]-1,2-dihydropyridin-2-one

  • MDL number MFCD01568913
  • Cas Number : 338393-87-6
  • Molecular Weight : 390.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65260

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1P-712_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart