KEY465193362 Key Organics / BIONET

1-[(2H-1,3-benzodioxol-5-yl)methyl]-3-[(2-chloro-6-fluorophenyl)methyl]-4-hydroxy-1,2-dihydropyridin-2-one

  • MDL number MFCD01871463
  • Cas Number : 477869-82-2
  • Molecular Weight : 387.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65261

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1P-713_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart