KEY465193364 Key Organics / BIONET

ethyl 2-{2-amino-[1,2,4]triazolo[1,5-a]pyrimidin-7-yl}-3-phenylpropanoate

  • MDL number MFCD02082380
  • Cas Number : 477869-91-3
  • Molecular Weight : 311.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65271

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1P-731_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart