KEY465193371 Key Organics / BIONET

4-{6-acetyl-5-methyl-4H,7H-[1,2,4]triazolo[1,5-a]pyrimidin-7-yl}benzoic acid

  • MDL number MFCD02082387
  • Cas Number : 477870-10-3
  • Molecular Weight : 298.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65302

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1P-828_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart