KEY465193375 Key Organics / BIONET

ethyl 2-[2-methyl-4-(4-nitrophenyl)-3-oxo-1,2,4-oxadiazolidin-5-yl]acetate

  • MDL number MFCD02082388
  • Cas Number : 477870-12-5
  • Molecular Weight : 309.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65309

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1P-836_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart