KEY465193386 Key Organics / BIONET

(E)-[(2,6-dichlorophenyl)methoxy]({[4-(3-fluoropropoxy)phenyl]methylidene})amine

  • MDL number MFCD02102353
  • Cas Number : 477870-27-2
  • Molecular Weight : 356.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65325

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1R-0021_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart