KEY465193396 Key Organics / BIONET

N-cyclopentyl-4-(2,6-dichlorobenzoyl)-1H-pyrrole-2-carboxamide

  • MDL number MFCD02102365
  • Cas Number : 477870-41-0
  • Molecular Weight : 351.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65344

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1R-0041_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart