KEY465193402 Key Organics / BIONET

N-[3-(dimethylamino)propyl]-4-(2-phenylacetyl)-1H-pyrrole-2-carboxamide

  • MDL number MFCD02570864
  • Cas Number : 477870-48-7
  • Molecular Weight : 313.401

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65350

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/1R-0047_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart