KEY465193405 Key Organics / BIONET

4,5-dimethylcyclohexa-1,4-diene-1,2-dicarboxylic acid

  • MDL number MFCD02185959
  • Cas Number : 91061-82-4
  • Molecular Weight : 196.202

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65357

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1R-0056_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart