KEY465193416 Key Organics / BIONET

(2E)-3-(dimethylamino)-2-[(E)-3,4,5-trimethyl-1H-pyrazole-1-carbonyl]prop-2-enenitrile

  • MDL number MFCD02102381
  • Cas Number : 477710-15-9
  • Molecular Weight : 232.287

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65382

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1R-0096_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart