KEY465193418 Key Organics / BIONET

(E)-2-(3,5-dimethyl-4-propyl-1H-pyrazol-1-yl)-N-(2-nitrophenyl)-2-oxoethanecarbohydrazonoyl cyanide

  • MDL number MFCD02102383
  • Cas Number : 477710-16-0
  • Molecular Weight : 354.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65386

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1R-0102_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart