KEY465193424 Key Organics / BIONET

(E)-N-(2-nitrophenyl)-2-oxo-2-(3,4,5-trimethyl-1H-pyrazol-1-yl)ethanecarbohydrazonoyl cyanide

  • MDL number MFCD02102388
  • Cas Number : 477710-19-3
  • Molecular Weight : 326.316

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65399

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1R-0122_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart