KEY465193425 Key Organics / BIONET

(E)-N-(2-nitrophenyl)-1-{N'-[(2E)-5,5,5-trifluoro-4-oxopentan-2-ylidene]hydrazinecarbonyl}methanecarbohydrazonoyl cyanide

  • MDL number MFCD02102390
  • Cas Number : 477870-87-4
  • Molecular Weight : 384.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65403

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1R-0131_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart